Quinidine sulfate dihydrate, 98+%, Thermo Scientific Chemicals
Quinidine sulfate dihydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Quinidine sulfate dihydrate, 98+%, Thermo Scientific Chemicals

A sodium channel blocker, and to a lesser extent also blocks potassium channels
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J60426.06
also known as J60426-06
Price (USD)/ Each
58.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
58.50
Add to cart
Quinidine sulfate dihydrate, 98+%, Thermo Scientific Chemicals
Catalog numberJ60426.06
Price (USD)/ Each
58.50
-
Add to cart
Chemical Identifiers
CAS6591-63-5
IUPAC Namebis((S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular FormulaC40H54N4O10S
InChI KeyZHNFLHYOFXQIOW-YKSBFIEBSA-N
SMILESO.O.OS(O)(=O)=O.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition≤0.2%
Appearance (Color)White
FormPowder
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥98.0%
View more

Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.

Solubility
Soluble in chloroform, water (11 mg/ml at 20°C), alcohol, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away fom strong oxidizing agents.
RUO – Research Use Only
ZK Shihabi. Organic solvent high-field amplified stacking for basic compounds in capillary electrophoresis. Journal of Chromatography A. 2005, 1066  ,(1-2), 205-210.