Thermo Scientific Chemicals

Myricetin, 98%, Thermo Scientific Chemicals

Catalog number: J60450.MA
10 mg, Each
Thermo Scientific Chemicals

Myricetin, 98%, Thermo Scientific Chemicals

Catalog number: J60450.MA
10 mg, Each
Quantity
Catalog number: J60450.MA
also known as J60450-MA
Price (USD)
Quantity
-
Have Questions?

Description

Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol. Insoluble in water.  

Notes
Store in a cool place. Incompatible with strong oxidizing agents.    
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text