Thermo Scientific Chemicals

Ipratropium bromide, 98%, Thermo Scientific Chemicals

Catalog number: J60453.MC
100 mg, Each
Thermo Scientific Chemicals

Ipratropium bromide, 98%, Thermo Scientific Chemicals

Catalog number: J60453.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60453.MC
also known as J60453-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22254-24-6
Assay (HPLC)
≥ 98 %
GC/MS Analysis
Conforms to structure
Melting Point
235 to 240?C
Appearance (Color)
White to cream
Form
Powder

Description

Muscarinic acetylcholine receptor antagonist and a bronchodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text