Amiodarone hydrochloride, Thermo Scientific Chemicals
Amiodarone hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amiodarone hydrochloride, Thermo Scientific Chemicals

A non-selective ion channel blocker; also an antiarrhythmic agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60456.14
also known as J60456-14
Price (USD)/ Each
157.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
157.00
Add to cart
Amiodarone hydrochloride, Thermo Scientific Chemicals
Catalog numberJ60456.14
Price (USD)/ Each
157.00
-
Add to cart
Specifications
Chemical Name or MaterialAmiodarone hydrochloride
CAS19774-82-4
Health Hazard 1H312+H332-H361d-H373
Health Hazard 3P201-P202-P260-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c
Melting Point156°C to 163°C
View more
Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Yong-Qiang Zhang; Rajini Rao. Global disruption of cell cycle progression and nutrient response by the antifungal agent amiodarone. Journal of Biological Chemistry. 2007, 282, (52), 37844-37853.
  2. Minhui Wang; Jiaojiao Shan; Qian Yang; Xianglei Ma; Sisi Jin; Xiaoke Guo; Qidong You; Yiqun Tang. Antiarrhythmic efficacy of CPUY102122, a multiple ion channel blocker, on rabbits with ischemia/reperfusion injury. Pharmacological reports : PR.2014, 66, (6), 1022-1030.