Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity, Thermo Scientific Chemicals

Catalog number: J60461.AP
500mL, Each
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity, Thermo Scientific Chemicals

Catalog number: J60461.AP
500mL, Each
Quantity
Catalog number: J60461.AP
also known as J60461-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54827-17-7
IUPAC Name
3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Formula
C16H20N2
InChI Key
UAIUNKRWKOVEES-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
Appearance (Color)
Colorless
Form
Liquid
Assay (special)
Free of Precipitate: Passes Test
Assay (special)
Dot Blot: Passes Test
Clarity
Transparent

Description

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text