Hexamethonium bromide, 98+%, Thermo Scientific Chemicals
Hexamethonium bromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexamethonium bromide, 98+%, Thermo Scientific Chemicals

Nicotinic acetyl choline receptor antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number J60466.22
also known as J60466-22
Price (USD)/ Each
208.65
Online exclusive
232.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
208.65
Online exclusive
232.00 
Save 23.35 (10%)
Add to cart
Hexamethonium bromide, 98+%, Thermo Scientific Chemicals
Catalog numberJ60466.22
Price (USD)/ Each
208.65
Online exclusive
232.00 
Save 23.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialHexamethonium bromide
CAS55-97-0
Melting Point282°C to 285°C (decomposition)
ColorWhite
Recommended StorageAmbient temperatures
View more
Nicotinic acetyl choline receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only