BSA Blocking Buffer, 3% in PBS, with 0.02% sodium azide
This product is animal origin and can only be shipped within the United States
BSA Blocking Buffer, 3% in PBS, with 0.02% sodium azide
Thermo Scientific Chemicals

BSA Blocking Buffer, 3% in PBS, with 0.02% sodium azide

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60473.AK
also known as J60473-AK
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
Add to cart
BSA Blocking Buffer, 3% in PBS, with 0.02% sodium azide
Catalog numberJ60473.AK
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
-
Add to cart
Specifications
Product TypeBlocking Buffer
Quantity250 mL
FormLiquid
Recommended StorageStore at -20°C
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C