Thermo Scientific Chemicals
Meclofenamic acid sodium salt, 99+%, Thermo Scientific Chemicals
Catalog number: J60484.03
1 g, Each
Thermo Scientific Chemicals
Meclofenamic acid sodium salt, 99+%, Thermo Scientific Chemicals
Catalog number: J60484.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60484.03
also known as J60484-03
Price (USD)
Quantity
-
Have Questions?
Description
Inhibits 5-lipoxygenase and cyclooxygenase; NSAID
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Meclofenamic acid is an aminobenzoic acid-based NSAID that is characterized by two aryl moieties
• Meclofenamic acid is an inhibitor of arachidonate 5-lipoxygenase and prostaglandin-endoperoxide synthase
• Meclofenamic acid prevents pyrogen release and inhibits wound granulation; it may also act as a neoplastic
Application
• Meclofenamic acid can complex metals, making it of interest to drug delivery studies that target its toxic metal complexes to cancerous cell types
• It is suitable for laboratory research on pain, degeneration, wound healing, and tumor growth