Meclofenamic acid sodium salt, 99+%
Meclofenamic acid sodium salt, 99+%
Meclofenamic acid sodium salt, 99+%
Meclofenamic acid sodium salt, 99+%
Thermo Scientific Chemicals

Meclofenamic acid sodium salt, 99+%

Meclofenamic acid (meclofenamate, CAS # 644-62-2) is a nonsteroidal anti-inflammatory drug (NSAID) with antipyretic and antineoplastic activities. | CAS: 6385-02-0 | C14H13Cl2NNaO2 | 321.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60484.06
also known as J60484-06
Price (USD)
191.00
Each
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
191.00
Each
Add to cart
Specifications
CAS6385-02-0
Chemical Name or MaterialMeclofenamic acid sodium salt
ColorWhite
EINECS Number228-983-3
Formula Weight318.13
View more
Ask our AI about this Product
Inhibits 5-lipoxygenase and cyclooxygenase; NSAID

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Meclofenamic acid is an aminobenzoic acid-based NSAID that is characterized by two aryl moieties

• Meclofenamic acid is an inhibitor of arachidonate 5-lipoxygenase and prostaglandin-endoperoxide synthase

• Meclofenamic acid prevents pyrogen release and inhibits wound granulation; it may also act as a neoplastic

Application

• Meclofenamic acid can complex metals, making it of interest to drug delivery studies that target its toxic metal complexes to cancerous cell types

• It is suitable for laboratory research on pain, degeneration, wound healing, and tumor growth

RUO – Research Use Only

General References:

  1. Soriano-Hernández AD, Galvan-Salazar HR, Montes-Galindo DA, Rodriguez-Hernandez A, Martinez-Martinez R, Guzman-Esquivel J, Valdez-Velazquez LL, Baltazar-Rodriguez LM, Espinoza-Gómez F, Rojas-Martinez A, Ortiz-Lopez R, Gonzalez-Alvarez R, Delgado-Enciso I. Antitumor effect of meclofenamic acid on human androgen-independent prostate cancer: a preclinical evaluation. Int Urol Nephrol. 2012 Apr;44(2):471-7. doi: 10.1007/s11255-011-0012-0. Epub 2011 Jun 10. PMID: 21660425.
  2. Saigo C, Nomura Y, Yamamoto Y, Sagata M, Matsunaga R, Jono H, Nishi K, Saito H. Meclofenamate elicits a nephropreventing effect in a rat model of ischemic acute kidney injury by suppressing indoxyl sulfate production and restoring renal organic anion transporters. Drug Des Devel Ther. 2014 Aug 13;8:1073-82. doi: 10.2147/DDDT.S67456. PMID: 25143712; PMCID: PMC4137993.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link