Thermo Scientific Chemicals

Acrylamide/Bisacrylamide 19:1, powder

Catalog number: J60486.22
100 g, Each
Thermo Scientific Chemicals

Acrylamide/Bisacrylamide 19:1, powder

Catalog number: J60486.22
100 g, Each
Quantity
Catalog number: J60486.22
also known as J60486-22
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
White
Assay from Supplier's CofA
Conductivity: ≤10µMho
Form
Powder

Description

Premixed powder for preparing protein PAGE gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text