Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals
LB Agar plate with 25μg/mL Kanamycin
Catalog number: J60487.EQF
20 Plates, Each
Thermo Scientific Chemicals
LB Agar plate with 25μg/mL Kanamycin
Catalog number: J60487.EQF
20 Plates, Each
Quantity
Catalog number: J60487.EQF
also known as J60487-=F
Price (USD)
Have Questions?
Description
Suitable for selective cultivation of E. coli strains that contain plasmids conferring kanamycin resistance. The bacteria can be used for DNA plasmid production or production of recombinant proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used for growth of Escherichia coli, plasmid and other bacterial cultures