Thermo Scientific Chemicals

Niflumic acid, 99+%, Thermo Scientific Chemicals

Catalog number: J60489.09
10 g, Each
Thermo Scientific Chemicals

Niflumic acid, 99+%, Thermo Scientific Chemicals

Catalog number: J60489.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60489.09
also known as J60489-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4394-00-7
IUPAC Name
2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
Molecular Formula
C13H9F3N2O2
InChI Key
JZFPYUNJRRFVQU-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F
Form
Powder or crystals or crystalline powder
Infrared spectrum
Conforms
Purity
≥99.0%

Description

It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

Solubility
Soluble in ethanol (∼50 mg/ml), acetone (50 mg/ml, Clear to Slightly Hazy, Yellow), methanol (∼50 mg/ml), DMSO (56 mg/ml at 25°C), and acetonitrile (∼50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Hygroscopic. Keep away fom strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text