Gel loading dye used in molecular biology protocolsThe 6X Orange DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is recommended for the analysis of small DNA molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Gel loading dye used in molecular biology protocolsThe 6X Orange DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is recommended for the analysis of small DNA molecules.
Solubility
Soluble in water.
Notes
Contains: 0.25% Orange-G and 30% glycerol in autoclaved DI water
RUO – Research Use Only
RJ Williams. Isolation and characterization of an unknown restriction endonuclease. Nuclease Methods and Protocols. 2001, 160, (1), 431-442.
PA Asare.; IKA Galyuon.; JK Sarfo.; JP Tetteh. Morphological and molecular based diversity studies of some cassava (Manihot esculenta crantz) germplasm in Ghana. African journal of biotechnology. 2013, 63, (10), 891-895.