Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Hyaluronic acid, bovine vitreous humor, Thermo Scientific Chemicals

Catalog number: J60566.MA
10 mg, Each
Thermo Scientific Chemicals

Hyaluronic acid, bovine vitreous humor, Thermo Scientific Chemicals

Catalog number: J60566.MA
10 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60566.MA
also known as J60566-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-61-9
IUPAC Name
sodium (2S,3S,4R,5R,6R)-3-{[(2S,3R,4R,5S,6R)-4-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3-acetamido-5-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-{[(2R,3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4,5-dihydroxyoxane-2-carboxylic acid
Molecular Formula
C28H44N2NaO23
InChI Key
YWIVKILSMZOHHF-QJZPQSOGSA-N
SMILES
[Na+].CC(=O)N[C@H]1[C@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@@H]([C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)[C@H]2NC(C)=O)[C@H](O)[C@H]1O)C(O)=O
Appearance (Color)
White to cream
Comment
Substrate suitability: Satisfactory for hyaluronidase assay
Form
Powder

Description

Hyaluronic acid is a naturally derived, non - immunogenic, non - adhesive glycosaminoglycan that plays a prominent role in various wound - healing processes, as it as it is naturally angiogenic when degraded to small fragments. Hyaluronic acid promotes early inflammation which is critical for initiating wound healing, but then moderates later stages of the process, allowing matrix stabilization and reduction of long term inflammation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Hyaluronic acid, derived from bovine vitreous humor, is a naturally derived, non-immunogenic, non-adhesive glycosaminoglycan that shows an angiogenic activity promoting wound-healing processes when degraded to small fragments
Application
<
• It is considered a mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine, which forms the core of complex proteoglycan aggregates found in the extracellular matrix
• It has been used to study the activation of the MAPK/ERK pathway following infection of macrophages with Leishmania amazonensis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text