Fast Garnet GBC base, Thermo Scientific Chemicals
Fast Garnet GBC base, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fast Garnet GBC base, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number J60574.22
also known as J60574-22
Price (USD)/ Each
26.40
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
26.40
Add to cart
Fast Garnet GBC base, Thermo Scientific Chemicals
Catalog numberJ60574.22
Price (USD)/ Each
26.40
-
Add to cart
Chemical Identifiers
CAS97-56-3
IUPAC Name2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
Molecular FormulaC14H15N3
InChI KeyPFRYFZZSECNQOL-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1N)N=NC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.0% min.
Melting Point100-103?
Loss on Drying3.0% max.
C.I. 11160

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 11160
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only