Lecithin, 60%, egg, Thermo Scientific Chemicals
Lecithin, 60%, egg, Thermo Scientific Chemicals
Lecithin, 60%, egg, Thermo Scientific Chemicals
Lecithin, 60%, egg, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lecithin, 60%, egg, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number J60576.14
also known as J60576-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS8002-43-5
IUPAC Name(2-{[3-(hexadecanoyloxy)-2-(octadeca-9,12-dienoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular FormulaC42H80NO8P
InChI KeyJLPULHDHAOZNQI-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Assay (special)≥ 97.0 % (as soluble in ethanol)
Loss on Drying≤ 1.5 %
Acid Value≤ 30 mg KOH/gm
Peroxide content≤ 12 meq O2/kg
View more
Lecithin is used as an emulsifying and stabilizing agent in food, pharmaceutical and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacture of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lecithin is used as an emulsifying and stabilizing agent in food, pharmaceutical and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacture of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

Solubility
Soluble in diethyl ether, absolute alcohol, chloroform and petroleum ether. Sparingly soluble in benzene. Insoluble in water and acetone .

Notes
Light, heat, moisture and air-sensitive. Do not store in direct sunlight. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Paz, P.; Esteso, M. C.; Alvarez, M.; Mata, M.; Charorro, C. A.; Anel, L. Development of extender based on soybean lecithin for its application in liquid ram semen. Theriogenology 2010, 74 (4), 663-671.
  2. Cabezas, D. M.; Diehl, B. W. K.; Tomas, M. C. Sunflower Lecithin: Application of a Fractionation Process with Absolute Ethanol. J. Am. Oil Chemists' Soc. 2009, 86 (2), 189-196.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection