Thermo Scientific Chemicals

CHAPS lysis buffer

Catalog number: J60580.AE
100 mL, Each
Thermo Scientific Chemicals

CHAPS lysis buffer

Catalog number: J60580.AE
100 mL, Each
Quantity
Catalog number: J60580.AE
also known as J60580-AE
Price (USD)
Have Questions?
Appearance (Color)
Pale yellow
Form
Liquid
Comment
Filtered through a 0.22 micron filter.
pH
7.4 ± 0.20
Purity
Raw material: ≥98.0%

Description

For cell lysis and protein extraction

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text