Sulforhodamine 101 acid chloride, Thermo Scientific Chemicals
Sulforhodamine 101 acid chloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulforhodamine 101 acid chloride, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 mg
Catalog number J60581.MA
also known as J60581-MA
Price (USD)/ Each
225.65
Online exclusive
251.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
225.65
Online exclusive
251.00 
Save 25.35 (10%)
Add to cart
Sulforhodamine 101 acid chloride, Thermo Scientific Chemicals
Catalog numberJ60581.MA
Price (USD)/ Each
225.65
Online exclusive
251.00 
Save 25.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialSulforhodamine 101 acid chloride
CAS82354-19-6
Health Hazard 1H301-H332-H351
Health Hazard 2GHS H Statement
H351-H373-H302
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c
View more
Fluorescent labeling compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only