Thermo Scientific Chemicals

Kemptide

Catalog number: J60591.LB0
5 mg, Synthetic peptide substrate for protein kinase A, Each
Thermo Scientific Chemicals

Kemptide

Catalog number: J60591.LB0
5 mg, Synthetic peptide substrate for protein kinase A, Each
Quantity
For Use With (Application)
Catalog number: J60591.LB0
also known as J60591-#0
Price (USD)
Quantity
-
Have Questions?
Form
Lyophilized Powder
Appearance (Color)
White
Purity
≥95.0%
Molecular Weight
Conforms with theoretical MW

Description

Kemptide, phosphate acceptor, is shown to be a basic serine-containing heptapeptide corresponding to a sequence from pig liver pyruvate kinase. This synthetic peptide is a substrate of PKA (cyclic AMP-dependent protein kinase) and is phosphorylated and activated after microinjection into Xenopus oocytes. Exposure of Kemptide to insulin has demonstrated an increase in activation of this agent, which can be inhibited by the catalytic subunit of phosphatase 2A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text