Thermo Scientific™

1,3-Dihydroxyacetone dimer, Thermo Scientific™

Catalog number: J60592.06
5 g, Each
Thermo Scientific™

1,3-Dihydroxyacetone dimer, Thermo Scientific™

Catalog number: J60592.06
5 g, Each
Quantity
Catalog number: J60592.06
also known as J60592-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62147-49-3
Appearance (Color)
White to pale cream
Form
Powder
Assay from Suppliers CofA
≥96.0%

Description

1,3-Dihydroxyacetone dimer is primarily used as an ingredient in sunless tanning products. It is a waterproof skin dye used to darken light or unpigmented areas of skin affected by vitiligo, scars or other causes.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-Dihydroxyacetone dimer is primarily used as an ingredient in sunless tanning products. It is a waterproof skin dye used to darken light or unpigmented areas of skin affected by vitiligo, scars or other causes.

Solubili
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text