Thermo Scientific Chemicals

Carrageenan, iota type, Thermo Scientific Chemicals

Catalog number: J60603.22
100 g, Each
Thermo Scientific Chemicals

Carrageenan, iota type, Thermo Scientific Chemicals

Catalog number: J60603.22
100 g, Each
Quantity
Catalog number: J60603.22
also known as J60603-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9062-07-1
Molecular Formula
(C12H16O15S2)n
SMILES
OC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](OS(O)(=O)=O)[C@@H](-*)O3)[C@H](O)[C@@H](O-*)[C@H]1OS(O)(=O)=O
MDL Number
MFCD00151512
Loss on Drying
≤12.5%
pH
8.0 - 11.0 (1% solution in water)
Appearance (Color)
White to pale cream
Form
Powder

Description

Carrageenan, iota type is used as a stabilizer, for suspending cocoa in chocolate manufacture, and to clarify beverages. Carrageenans are used commercially as thickeners and stabilizing agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carrageenan, iota type is used as a stabilizer, for suspending cocoa in chocolate manufacture, and to clarify beverages. Carrageenans are used commercially as thickeners and stabilizing agents.

Solubility
Partially soluble in water, Soluble in hot water. Insoluble in ethanol, in hot concentrated salt solutions.

Notes
Stable under normal conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text