Thermo Scientific Chemicals

Meloxicam, Thermo Scientific Chemicals

Catalog number: J60635.06
5 g, Each
Thermo Scientific Chemicals

Meloxicam, Thermo Scientific Chemicals

Catalog number: J60635.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60635.06
also known as J60635-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71125-38-7
IUPAC Name
(3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
Molecular Formula
C14H13N3O4S2
InChI Key
DWMREKMVXIFPFM-ACCUITESSA-N
SMILES
CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O
Appearance (Color)
Pale yellow to yellow or yellow-green
Assay from Supplier's CofA
≥97% (HPLC)
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Mass Spec
Conforms to structure

Description

Meloxicam is developed for the treatment of rheumatoid arthritis and osteoarthritis. It is currently for the treatment of osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (22 mg/ml), DMSO (25 mg/ml), DMF, acetone(slightly soluble), ethanol(slightly soluble), and methanol(slightly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text