Thermo Scientific Chemicals

DNAzol™ Reagent

Catalog number: J60637.AD
50 mL, Each
Thermo Scientific Chemicals

DNAzol™ Reagent

Catalog number: J60637.AD
50 mL, Each
Quantity
Catalog number: J60637.AD
also known as J60637-AD
Price (USD)
Have Questions?
Appearance (Color)
Blue-green
Form
Slightly viscous liquid
Assay (special)
Integrity and sutability of DNA tested by PCR: Passes test
Assay (special)
Long PCR: Tested in generating multiple bands of 20 to 100 kb fragments
Assay (special)
Restriction enzyme digest: Passes test

Description

DNAzol™ Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
  • Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
  • A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
  • It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems

Applications

  • Suitable for in vitro and in vivo applications
  • Stabilized in powder form for laboratory use only
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text