Thermo Scientific Chemicals

Adenosine-5'-diphosphate monopotassium salt dihydrate, 99%

Catalog number: J60672.03
1 g, Each
Thermo Scientific Chemicals

Adenosine-5'-diphosphate monopotassium salt dihydrate, 99%

Catalog number: J60672.03
1 g, Each
Quantity
Catalog number: J60672.03
also known as J60672-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72696-48-1
IUPAC Name
potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-({[(hydrogen phosphonatooxy)(hydroxy)phosphoryl]oxy}methyl)oxolane-3,4-diol
Molecular Formula
C10H14KN5O10P2
InChI Key
ZNCWUOPIJTUALR-GWKNMROSNA-M
SMILES
[K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Form
Powder
Appearance (Color)
White to off-white
Assay from Suppliers CofA
≥98.5% (HPLC)
Solution Test
Clarity & Color: Clear and colorless
Loss on Drying
5.0-8.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text