4-Thiouridine, 98+%, Thermo Scientific Chemicals
4-Thiouridine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Thiouridine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
100 mg
Catalog number J60679.MD
also known as J60679-MD
Price (USD)/ Each
674.65
Online exclusive
749.00 
Save 74.35 (10%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
674.65
Online exclusive
749.00 
Save 74.35 (10%)
Add to cart
4-Thiouridine, 98+%, Thermo Scientific Chemicals
Catalog numberJ60679.MD
Price (USD)/ Each
674.65
Online exclusive
749.00 
Save 74.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13957-31-8
SpecificationsSpecification SheetSpecification Sheet
Purity≥98.0%
Appearance (Color)Off-white to pale yellow
FormPowder
4-Thiouridine is a constituent of several transfer RNA's. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Thiouridine is a constituent of several transfer RNA′s. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

Solubility
Soluble in water (20 mg/ml), and methanol (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

General References:

  1. A. Favre,; M. Yaniv,; A.M. Michels.The photochemistry of 4-thiouridine in escherichia coli t-RNAİVal. Biochemical and Biophysical Research Communications. 1969, 37 (2), 266-271.
  2. L. Aravind,; Eugene V. Koonin.THUMP - a predicted RNA-binding domain shared by 4-thiouridine, pseudouridine synthases and RNA methylases. Trends in biochemical science. 2001, 26 (4) ,215-217.