Thermo Scientific Chemicals

alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals

Catalog number: J60687.06
5 g, Each
Thermo Scientific Chemicals

alpha-Cyclodextrin, 97+%, Thermo Scientific Chemicals

Catalog number: J60687.06
5 g, Each
Quantity
Catalog number: J60687.06
also known as J60687-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10016-20-3
IUPAC Name
5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
Molecular Formula
C36H60O30
InChI Key
HFHDHCJBZVLPGP-UHFFFAOYNA-N
SMILES
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Assay (Titration)
≥ 97.0 %
Impurity content
Organic volatile impurities: ≤ 20 ppm
Appearance (Color)
White to pale cream
Residue on ignition
≤ 0.1 %
Water Content (Karl Fischer Titration)
≤ 10 %

Description

Useful for selective precipitation of enantiomeric, positional or structural isomersα-Cyclodextrin is used as a fiber ingredient, an odor or flavor masking agent. It is also useful for emulsification applications. It is also used as whipping fiber and emulsifying fiber. It finds application in medical, healthcare and food and beverage applications. It is also used to lower blood low-density lipoprotein cholesterol levels and lower blood triglyceride levels. It plays an essential role in fat free or fat containing dessert compositions and also employed for the reduction or the replacement of egg white in confectionary and bakery applications. Further, it acts as a supramolecular carrier, complexing agent and controlled drug release. In addition to this, it is used to increase the insulin and leptin sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 1%(w/v)

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text