Thermo Scientific Chemicals

Ethambutol dihydrochloride, Thermo Scientific Chemicals

Catalog number: J60695.06
5 g, Each
Thermo Scientific Chemicals

Ethambutol dihydrochloride, Thermo Scientific Chemicals

Catalog number: J60695.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60695.06
also known as J60695-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1070-11-7
IUPAC Name
dihydrogen (2S)-2-[(2-{[(2S)-1-hydroxybutan-2-yl]amino}ethyl)amino]butan-1-ol dichloride
Molecular Formula
C10H26Cl2N2O2
InChI Key
AUAHHJJRFHRVPV-BZDVOYDHSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].CC[C@@H](CO)NCCN[C@@H](CC)CO
Loss on Drying
0.5% max.
Assay (unspecified)
98.0-100.5%
Assay (unspecified)
Specific rotation: +6.0 to +6.7 (c=10 in water at 25?)
Trace Metals
20 ppm max.

Description

Ethambutol dihydrochloride it acts as a tuberculostatic antibacterial agent. It is used to treat tuberculosis in combination with other antibiotics to reduce the development of drug resistance. It has been used to study multidrug-resistant tuberculosis (MDR-TB) in highest-incidence-rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text