Thermo Scientific Chemicals

HEPES, 1.0M buffer soln., pH 7.5

Catalog number: J60712.AK
250 mL, Each
Thermo Scientific Chemicals

HEPES, 1.0M buffer soln., pH 7.5

Catalog number: J60712.AK
250 mL, Each
Quantity
Catalog number: J60712.AK
also known as J60712-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7365-45-9
IUPAC Name
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Formula
C8H18N2O4S
InChI Key
JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES
OCCN1CCN(CCS(O)(=O)=O)CC1
Form
Liquid
pH
7.5±0.2
Appearance (Color)
Clear colorless
Comment
Components is dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 0.22 micron filter.

Description

HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.              
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text