Thermo Scientific Chemicals

Ouabain octahydrate, 95%, Thermo Scientific Chemicals

Catalog number: J60724.03
1 g, Each
Thermo Scientific Chemicals

Ouabain octahydrate, 95%, Thermo Scientific Chemicals

Catalog number: J60724.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60724.03
also known as J60724-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11018-89-6
IUPAC Name
4-[(1R,7S,9R,9bS,10R)-3a,5a,9,10-tetrahydroxy-9a-(hydroxymethyl)-11a-methyl-7-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one octahydrate
Molecular Formula
C29H60O20
InChI Key
TYBARJRCFHUHSN-HMNHGHQSSA-N
SMILES
O.O.O.O.O.O.O.O.CC1OC(O[C@H]2C[C@@H](O)C3(CO)[C@H]4[C@H](O)CC5(C)[C@H](CCC5(O)C4CCC3(O)C2)C2=CC(=O)OC2)C(O)C(O)C1O
Appearance (Color)
White to off-white to pale yellow
Form
Powder
Assay (UV/vis)
≥94.0%
Loss on Drying
14.5-22.0%

Description

Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

Solubility
Soluble in water.

Notes
Protect from light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text