Thermo Scientific Chemicals

(-)-Gallocatechin, Thermo Scientific Chemicals

Catalog number: J60731.MA
10 mg, Each
Thermo Scientific Chemicals

(-)-Gallocatechin, Thermo Scientific Chemicals

Catalog number: J60731.MA
10 mg, Each
Quantity
Catalog number: J60731.MA
also known as J60731-MA
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3371-27-5
IUPAC Name
2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol
Molecular Formula
C15H14O7
InChI Key
XMOCLSLCDHWDHP-UHFFFAOYNA-N
SMILES
OC1CC2=C(O)C=C(O)C=C2OC1C1=CC(O)=C(O)C(O)=C1
Mass Spec
M-H+: 305.8
Appearance (Color)
White to cream to brown
Identification (FTIR)
Conforms
Assay from Supplier's CofA
≥98.0% (LC)
lambda-max (UV-Vis)
205-215nm

Description

Polyphenolic compound from green tea. Possesses free-radical scavenging ability

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polyphenolic compound from green tea
  • Possesses free-radical scavenging ability
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text