TE buffer, (20X), pH 7.4, autoclaved
TE buffer, (20X), pH 7.4, autoclaved
Thermo Scientific Chemicals

TE buffer, (20X), pH 7.4, autoclaved

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number J60738.AP
also known as J60738-AP
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
Add to cart
TE buffer, (20X), pH 7.4, autoclaved
Catalog numberJ60738.AP
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialTE buffer
CAS1185-53-1
Recommended StorageAmbient temperatures
EINECS Number231-791-2
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only