ACES Buffer (0.5 M, pH 7.0)
ACES Buffer  (0.5 M, pH 7.0)
Thermo Scientific Chemicals

ACES Buffer (0.5 M, pH 7.0)

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number J60755.AK
also known as J60755-AK
Price (USD)/ Each
97.20
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
97.20
Add to cart
ACES Buffer (0.5 M, pH 7.0)
Catalog numberJ60755.AK
Price (USD)/ Each
97.20
-
Add to cart
Specifications
Chemical Name or MaterialACES buffer soln.
CAS7365-82-4
ColorColorless
Recommended StorageKeep cold
Concentration0.5 M
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only