Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X) pH 7.4

Catalog number: J60764.K7
4 L, Each
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X) pH 7.4

Catalog number: J60764.K7
4 L, Each
Quantity
Catalog number: J60764.K7
also known as J60764-K7
Price (USD)
Quantity
-
Have Questions?
Composition
250mM Tris-HCl, 27mM KCl, 1.37M NaCl
pH
7.4 ± 0.2
Form
Liquid
Comment
All components are dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 1 micron filter.
Appearance (Color)
Clear colorless

Description

TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.4.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text