Thermo Scientific Chemicals

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, Thermo Scientific Chemicals

Catalog number: J60767.AD
50 mL, Each
Thermo Scientific Chemicals

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, Thermo Scientific Chemicals

Catalog number: J60767.AD
50 mL, Each
Quantity
Catalog number: J60767.AD
also known as J60767-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-42-5
IUPAC Name
3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Molecular Formula
C14H24N2O10
InChI Key
DEFVIWRASFVYLL-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
Appearance (Color)
Clear colorless
Form
Liquid
Comment
Prepared in UV trated18.2megohm-cm ± 1 water, pH adjusted and filtered through a 0.22 micron filter.
pH
8.0 ± 0.20

Description

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol-O,O′-bis(2-aminoethyl)-N,N,N′,N′-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.

Solubility
Miscible with 0.1M sodium hydroxide solution.

Notes
Incompatible with strong oxidizing agents, alkali, alkali earth metal and many reactive organic and inorganic chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text