Resveratrol, 98%, Thermo Scientific Chemicals
Resveratrol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Resveratrol, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60790.ME
also known as J60790-ME
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
Add to cart
Resveratrol, 98%, Thermo Scientific Chemicals
Catalog numberJ60790.ME
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS501-36-0
IUPAC Name5-[(1E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Molecular FormulaC14H12O3
InChI KeyLUKBXSAWLPMMSZ-OWOJBTEDSA-N
SMILESOC1=CC=C(C=CC2=CC(O)=CC(O)=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Purity≥97.5%
It is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cells (MSC’s) in vitro. Resveratrol acts as a NF-κB inhibitor to protect MSC-derived chondrocytes in vitro. Resveratrol is used to treat adipose-derived mesenchymal stem cells (ADMSCs) resulted in increased alkaline phosphatase activity and osteocalcin levels; thus enriching the MSC and osteoprogenitor cell populations in cultured ADMSCs in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cel
RUO – Research Use Only

General References:

  1. Kimura et al. Effects of stilbenes on arachidonate metabolism in leukocytes. Biochim.Biophys.Acta . 1985, 834 ,(2), 275-8.
  2. Chun et al . Resveratrol is a selective human cytochrome P450 1A1 inhibitor. Biochem.Biophys.Res.Commun.1999, 262 ,(1), 20-4.