(+/-)-Miconazole nitrate, 98+%, Thermo Scientific Chemicals
(+/-)-Miconazole nitrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-Miconazole nitrate, 98+%, Thermo Scientific Chemicals

Inhibits cytochrome P450-dependent 14a-demethylase
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60796.06
also known as J60796-06
Price (USD)/ Each
117.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
117.00
Add to cart
(+/-)-Miconazole nitrate, 98+%, Thermo Scientific Chemicals
Catalog numberJ60796.06
Price (USD)/ Each
117.00
-
Add to cart
Specifications
Chemical Name or Material(+/-)-Miconazole nitrate
CAS22832-87-7
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
View more
Inhibits cytochrome P450-dependent 14a-demethylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only