Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot

Catalog number: J60801.K2
1 L, Each
Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot

Catalog number: J60801.K2
1 L, Each
Capacity
Catalog number: J60801.K2
also known as J60801-K2
Price (USD)
Quantity
-
Have Questions?
Comment
Prepared in in UV treated 18.2 megohm-cm ±1 water, followed by pH adjustment and filtered through 1 micron filtration.
Form
Liquid
pH
7.4 ± 0.2 (at 1X dilution)
Appearance (Color)
Clear colorless
Composition
1.5M NaCl and 100mM Sodium Phosphate

Description

Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and used to disengage attached and clumped cells. It also helps to maintain a constant pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text