Thermo Scientific Chemicals

Acrylamide/Bisacrylamide 29:1, powder

Catalog number: J60824.22
100 g, Each
Thermo Scientific Chemicals

Acrylamide/Bisacrylamide 29:1, powder

Catalog number: J60824.22
100 g, Each
Quantity
Catalog number: J60824.22
also known as J60824-22
Price (USD)
Quantity
-
Have Questions?
Assay (special)
Electrophoresis: Passes Test
Appearance (Color)
White
Solution Test
Solubility: Passes Test
Form
Solid

Description

Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

Solubility
It is soluble in water.

Notes
Keep away from light. Store at room tempeature.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text