Thermo Scientific Chemicals

Sodium phosphate, 0.5M buffer soln., pH 8.0

Catalog number: J60825.AP
500 mL, Each
Thermo Scientific Chemicals

Sodium phosphate, 0.5M buffer soln., pH 8.0

Catalog number: J60825.AP
500 mL, Each
Quantity
Catalog number: J60825.AP
also known as J60825-AP
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
pH
8.0 ± 0.5
Appearance (Color)
Clear colorless
Comment
Components are dissolved in UV treated 18.2 megohm-cm ± 1 water and filtered through 0.22 micron filter.

Description

Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text