Glucagon (1-29) trifluoroacetate salt, human
Glucagon (1-29) trifluoroacetate salt, human
Thermo Scientific Chemicals

Glucagon (1-29) trifluoroacetate salt, human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
0.5 mg
Catalog number J60827.MCR
also known as J60827-M^
Price (USD)/ Each
316.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
316.00
Add to cart
Glucagon (1-29) trifluoroacetate salt, human
Catalog numberJ60827.MCR
Price (USD)/ Each
316.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesHuman
Purification MethodHPLC
FormatPowder
For Use With (Application)A peptide hormone that plays a role in maintaining glucose homeostasis
Molecular Weight (g/mol)3482.74
View more
A peptide hormone that plays a role in maintaining glucose homeostasis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only