SSC buffer (20X)
Thermo Scientific Chemicals
SSC buffer (20X)
Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
2 L
Catalog number J60839.K2
also known as J60839-K2
Price (USD)/ Each
45.65
Online exclusive
50.30 
Save 4.65 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.30 
Save 4.65 (9%)
Add to cart
SSC buffer (20X)
Catalog numberJ60839.K2
Price (USD)/ Each
45.65
Online exclusive
50.30 
Save 4.65 (9%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Quantity1 L
Unit SizeEach
SSC buffer is suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures