Nitrendipine, Thermo Scientific Chemicals
Nitrendipine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitrendipine, Thermo Scientific Chemicals

L-type calcium channel blocker
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60841.03
also known as J60841-03
Price (USD)/ Each
660.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
660.00
Add to cart
Nitrendipine, Thermo Scientific Chemicals
Catalog numberJ60841.03
Price (USD)/ Each
660.00
-
Add to cart
Chemical Identifiers
CAS39562-70-4
IUPAC Name3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular FormulaC18H20N2O6
InChI KeyPVHUJELLJLJGLN-UHFFFAOYNA-N
SMILESCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA≥95.0%
Nitrendipine modulates NMDA receptor channel function in mammalian neurons. Nitrendipine has been shown to inhibit neutrophil adhesion to vascular endothelium. It is used in the treatment of primary hypertension to decrease blood pressure. Nitrendipine is used alone or with an angiotensin-converting enzyme inhibitor, to treat hypertension, chronic stable angina pectoris, and Prinzmetal's variant angina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO (17.5 mg/ml), methanol (15 mg/ml), ethyl acetate, and ethanol (25 mM). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. X Chen et. al.Quantitative determination of nitrendipine and its metabolite dehydronitrendipine in human plasma using liquid chromatography-tandem mass spectrometry.. Biomedical Chromatography. 2001, 15 (8), 518-524 .
  2. J Muntwyler and F Follath.Calcium channel blockers in treatment of hypertension.. Progress in Cardiovascular Diseases. 2001, 44 (3), 207-216.