Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 6.5

Catalog number: J60845.AK
250 mL, Each
Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 6.5

Catalog number: J60845.AK
250 mL, Each
Quantity
Catalog number: J60845.AK
also known as J60845-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7558-80-7
IUPAC Name
sodium dihydrogen phosphate
Molecular Formula
H2NaO4P
InChI Key
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES
[Na+].OP(O)([O-])=O
Composition
500mM Sodium Phosphate Monobasic
Comment
Prepared in 18.2 megohm - cm ± 1 water and filtered through 0.22 micron filter
Comment
Ultrapure Sodium Phosphate in given concentration dissolved in 18.2 megohm UV treated water, adjusted pH, filtered through 0.22 micron filter
pH
6.5 ± 0.15
Appearance (Color)
Clear, colorless liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text