Potassium phosphate, 0.2M buffer soln., pH 8.0
Potassium phosphate, 0.2M buffer soln., pH 8.0
Thermo Scientific Chemicals

Potassium phosphate, 0.2M buffer soln., pH 8.0

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J60851.AP
also known as J60851-AP
Price (USD)/ Each
94.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
94.00
Add to cart
Potassium phosphate, 0.2M buffer soln., pH 8.0
Catalog numberJ60851.AP
Price (USD)/ Each
94.00
-
Add to cart
Specifications
Chemical Name or MaterialPotassium phosphate
CAS7778-77-0
Recommended StorageAmbient temperatures
Concentration0.2 M
pH8.0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only