Thermo Scientific Chemicals

Miconazole, 97%, Thermo Scientific Chemicals

Catalog number: J60872.14
25 g, Each
Thermo Scientific Chemicals

Miconazole, 97%, Thermo Scientific Chemicals

Catalog number: J60872.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60872.14
also known as J60872-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22916-47-8
IUPAC Name
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
Molecular Formula
C18H14Cl4N2O
InChI Key
BYBLEWFAAKGYCD-UHFFFAOYNA-N
SMILES
ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
Melting Point
75.0-82.0°C
Appearance (Color)
White
Form
Powder
Assay from Supplier's CofA
≥96.0% (HPLC)

Description

Miconazole, is used as an antifungal inhibitor of aromatase. Miconazole has been shown to promote remyelination of neurons in chronic progressive multiple sclerosis mouse models. Miconazole is mainly used externally for the treatment of athlete's foot, ringworm, and jock itch. Internal application is used for oral or vaginal thrush (yeast infection). The oral gel may also be used for the lip disorder angular cheilitis. It is also used in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Freely soluble in alcohols or acetone. Also soluble in DMF or chloroform. Insoluble in water

Notes
Protect from light. Store in well-closed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text