Thermo Scientific Chemicals

Uricase, Candida utilis, Thermo Scientific Chemicals

Catalog number: J60875.EXR
100 U, Plastic bottle, Each
Thermo Scientific Chemicals

Uricase, Candida utilis, Thermo Scientific Chemicals

Catalog number: J60875.EXR
100 U, Plastic bottle, Each
Quantity
Packaging
Catalog number: J60875.EXR
also known as J60875-!R
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-12-4
IUPAC Name
{[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}({[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy})phosphinic acid amine
Molecular Formula
C18H26N5O14P
InChI Key
VPXDHZMLJOJKOX-UHFFFAOYNA-N
SMILES
N.OCC1OC(C(O)C1OP(O)(=O)OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O)N1C=CC(=O)NC1=O
Unit definition
One unit oxidizes 1 micromole of uric acid per minute at 25?C, pH 8.5
units/mgdw
2.0 minimum
UV/Vis
A260, 1mg/ml: Report value
UV/Vis
A280, 1mg/ml: Report value
Form
Lyophilized powder

Description

Uricase is very important for the determination of uric acid in biological fluids. Uricase is an important medical enzyme which can be used to determine urate in clinical analysis. The enzyme Urate oxidase (UO) catalyzes the oxidation of uric acid to 5-hydroxyisourate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text