Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 20X) pH 7.4

Catalog number: J60877.K2
1 L, Each
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 20X) pH 7.4

Catalog number: J60877.K2
1 L, Each
Quantity
Catalog number: J60877.K2
also known as J60877-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
Appearance (Color)
Clear colorless
Comment
Components are dissolved in UV treated 18.2 megohm-cm ± 1 water and filtered through 1 micron filter.
pH
7.4 ± 0.15
Form
Liquid

Description

Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

Solubility
It is soluble in water.

Notes
Contains: 0.5M Tris, 54mM KCl, and 2.74M NaCl, pH 7.4
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text