TRIS-buffered saline (TBS, 20X) pH 7.4
TRIS-buffered saline (TBS, 20X) pH 7.4
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 20X) pH 7.4

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 L
1 L
Catalog number J60877.K3
also known as J60877-K3
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
2 L
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
Add to cart
TRIS-buffered saline (TBS, 20X) pH 7.4
Catalog numberJ60877.K3
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
CommentComponents are dissolved in UV treated 18.2 megohm-cm ± 1 water and filtered through 1 micron filter.
pH7.4 ± 0.15
FormLiquid
Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

Solubility
It is soluble in water.

Notes
Contains: 0.5M Tris, 54mM KCl, and 2.74M NaCl, pH 7.4
RUO – Research Use Only
A Molloy.; P Laochumroonvorapong. Apoptosis, but not necrosis, of infected monocytes is coupled with killing of intracellular bacillus Calmette-Guerin. JEM. 1994, 180  ,(4), 1499-1509 .