Nicorandil, 98+%, Thermo Scientific Chemicals
Nicorandil, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nicorandil, 98+%, Thermo Scientific Chemicals

A vasodilator that activates ATP-sensitive potassium channels
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60879.03
also known as J60879-03
Price (USD)/ Each
306.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
306.00
Add to cart
Nicorandil, 98+%, Thermo Scientific Chemicals
Catalog numberJ60879.03
Price (USD)/ Each
306.00
-
Add to cart
Specifications
Chemical Name or MaterialNicorandil
CAS65141-46-0
Health Hazard 1H302-H318
Health Hazard 2GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
View more
A vasodilator that activates ATP-sensitive potassium channels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only