2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals
2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60927.MD
also known as J60927-MD
Price (USD)/ Each
180.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
180.00
Add to cart
2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals
Catalog numberJ60927.MD
Price (USD)/ Each
180.00
-
Add to cart
Specifications
Chemical Name or Material2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine
CAS123318-82-1
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A purine nucleoside anti-metabolite
  • Toxic to nondividing lymphocytes and monocytes
RUO – Research Use Only